Merck
Determination of lipoic acid by flow-injection and high-performance liquid chromatography with chemiluminescence detection.
Talanta (2012-07-24)
E Wołyniec, J Karpińska, S Losiewska, M Turkowicz, J Klimczuk, A Kojło
ABSTRAKT
A new flow-injection (FI) and high performance liquid chromatography (HPLC) with chemiluminescence detection method has been proposed for the determination of α-lipoic acid (LA). The assay is based on the measurement of chemiluminescence (CL) produced during the reaction of α-lipoic acid with potassium permanganate in a sodium hexametaphosphate medium (pH 3). This reaction is accompanied by a weak CL, which is greatly increased in the presence of a formaldehyde solution. The proposed FI method allows the determination of LA over the range: 0.5-20μgmL(-1) with LOD 4×10(-3)μgmL(-1). An introduction of HPLC into the flow manifold improves selectivity of the method and allows the determination of LA in a complex sample. The chromatographic linear range is 2.5-30μgmL(-1) with LOD 1.774μgmL(-1). Chromatographic separation was achieved by isocratic elution (acetonitrile/potassium dihydrogen phosphate, pH 3, adjusted with phosphoric acid): 30/70 using a Cosmosil 5C(18)-MS-II (4.6mm×150mm I.D.) column at a flow rate of 1.0mLmin(-1). The presented methods were utilized to determine the α-lipoic acid content in "Alfa-lipoic acid" capsules and in food products.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium permanganate, 97%
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%