Merck
Transesterification of diethyl oxalate with phenol over sol-gel MoO(3)/TiO(2) catalysts.
ChemSusChem (2012-07-24)
Trupti Kotbagi, Duy Luan Nguyen, Christine Lancelot, Carole Lamonier, Kaew-Arpha Thavornprasert, Zhu Wenli, Mickaël Capron, Louise Jalowiecki-Duhamel, Shubhangi Umbarkar, Mohan Dongare, Franck Dumeignil
ABSTRAKT
The transesterification of diethyl oxalate (DEO) with phenol to form diphenyl oxalate (DPO) has been carried out in the liquid phase over very efficient MoO(3)/TiO(2) solid-acid sol-gel catalysts. A selectivity of 100 % with a remarkable maximum yield of 88 % were obtained, which opens the route to downstream phosgene-free processes for the synthesis of polycarbonates. Interpretation of the results of various acidity measurements (NH(3) and pyridine desorption, methanol oxidation as a probe reaction) allowed us to identify the catalytic sites as Lewis acid sites.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Molybdenum(VI) oxide, 99.97% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, puriss. p.a., 99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, ACS reagent, ≥99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, nanopowder, 100 nm (TEM), 99.5% trace metals basis