Merck
Synthesis of phenserine analogues and evaluation of their cholinesterase inhibitory activities.
Bioorganic & medicinal chemistry (2012-07-27)
Masashi Shinada, Fuminori Narumi, Yuji Osada, Koji Matsumoto, Takayasu Yoshida, Kazuhiro Higuchi, Tomomi Kawasaki, Hiroyuki Tanaka, Mitsutoshi Satoh
ABSTRAKT
Phenserine is a potentially attractive drug for Alzheimer's disease. In order to further expand SAR study for inhibitions of acetylcholinesterase (AChE) and butyrylcholinesterase (BuChE), the methyl group at the 3a-position of phenserine was replaced with an alkyl or alkenyl group, and its phenylcarbamoyl moiety was substituted at the o- or p-position. The synthetic methodology for these phenserine analogues includes the efficient cascade reactions for introduction of the 3a-substituent and assembly of the quaternary carbon center followed by reductive cyclization to the key pyrroloindoline structure. The bulkiness of the substituent at 3a-position of phenserine derivatives tends to reduce the inhibitory effect on AChE activity in the following order: methyl > ethyl > vinyl > propyl ≈ allyl > reverse-prenyl groups. Among the series synthesized, the 3a-ethyl derivative demonstrated the highest AChE selectivity. In construct, the 3a-reverse-prenyl derivative indicated modest BuChE selectivity.