Merck
  • Synthesis of 3-(1-alkyl/aminoalkyl-3-vinyl-piperidin-4-yl)-1-(quinolin-4-yl)-propan-1-ones and their 2-methylene derivatives as potential spermicidal and microbicidal agents.
Synthesis of 3-(1-alkyl/aminoalkyl-3-vinyl-piperidin-4-yl)-1-(quinolin-4-yl)-propan-1-ones and their 2-methylene derivatives as potential spermicidal and microbicidal agents.
Bioorganic & medicinal chemistry letters (2012-08-01)
Rishi Ranjan Pandey, Akansha Srivastava, Richa Malasoni, Arshi Naqvi, Ashish Jain, Jagdamba Prasad Maikhuri, Sarvesh Paliwal, Gopal Gupta, Anil Kumar Dwivedi
ABSTRAKT
A series of twenty two derivatives of 3-(1-alkyl/aminoalkyl-3-vinyl-piperidin-4-yl)-1-(quinolin-4-yl)-propan-1-one and their 2-methylene derivatives were synthesized from naturally abundant cinchonine (I). Tartarate salts of these compounds were prepared and evaluated for spermicidal activity. The most active compounds (24, 27, 34, 36, and 38) showing potent spermicidal activity were further evaluated against different strains of Trichomonas vaginalis, for antimicrobial activity, in HeLa cell lines for cytotoxicity and against Lactobacillus jensenii for eco-safety. The tartarate of 3-(1-pentyl-3-vinyl-piperidin-4-yl)-1-(quinolin-4-yl)-propan-1-one (27) was found to be more active than N-9 in spermicidal activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cinchonine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
(+)-Cinchonine, 85%