Merck
A convenient ultrasound-promoted synthesis of some new thiazole derivatives bearing a coumarin nucleus and their cytotoxic activity.
Molecules (Basel, Switzerland) (2012-08-07)
Sobhi M Gomha, Khaled D Khalil
ABSTRAKT
Successful implementation of ultrasound irradiation for the rapid synthesis of a novel series of 3-[1-(4-substituted-5-(aryldiazenyl)thiazol-2-yl)hydrazono)ethyl]-2H-chromen-2-ones 5a-h, via reactions of 2-(1-(2-oxo-2H-chromen-3-yl)ethylidene) thiosemicarbazide (2) and the hydrazonoyl halides 3(4), was demonstrated. Also, a new series of 5-arylidene-2-(2-(1-(2-oxo-2H-chromen-3-yl)ethylidene)hydrazinyl)thiazol-4(5H)-ones 10a-d were synthesized from reaction of 2 with chloroacetic acid and different aldehydes. Moreover, reaction of 2-cyano-N'-(1-(2-oxo-2H-chromen-3-yl)ethylidene)-acetohydrazide (12) with substituted benzaldehydes gave the respective arylidene derivatives 13a-c under the conditions employed. The structures of the synthesized compounds were assigned based on elemental analyses and spectral data. Also, the cytototoxic activities of the thiazole derivative 5a was evaluated against HaCaT cells (human keratinocytes). It was found that compound 5a possess potent cytotoxic activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,4-Dioxane solution, suitable for NMR (reference standard), 40% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane solution, suitable for NMR (reference standard), 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane solution, suitable for NMR (reference standard), 10 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.
Supelco
1,4-Dioxane solution, suitable for NMR (reference standard), 40% in benzene-d6 (99.6 atom % D), NMR tube size 5 mm × 8 in.
Supelco
1,4-Dioxane solution, suitable for NMR (reference standard), 5 mM in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane solution, suitable for NMR (reference standard), 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane solution, suitable for NMR (reference standard), 40% in benzene-d6 (99.6 atom % D)
Supelco
1,4-Dioxane, analytical standard
Supelco
1,4-Dioxane solution, certified reference material, 2000 μg/mL in methanol, ampule of 1 mL
Supelco
1,4-Dioxane solution, suitable for NMR (reference standard), 5 mM in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane, suitable for HPLC, ≥99.5%
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
2-Bromoacetophenone, 98%
Sigma-Aldrich
Chloroacetone, Wacker Chemie AG, ≥96.0% (GC)
Supelco
2-Bromoacetophenone, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Sigma-Aldrich
Chloroacetone, 95%