Merck
VKORC1-dependent pharmacokinetics of intravenous and oral phylloquinone (vitamin K1) mixed micelles formulation.
European journal of clinical pharmacology (2012-08-07)
Milka Marinova, Dieter Lütjohann, Olof Breuer, Heike Kölsch, Philipp Westhofen, Matthias Watzka, Martin Mengel, Birgit Stoffel-Wagner, Gunther Hartmann, Christoph Coch, Johannes Oldenburg
ABSTRAKT
The pharmacokinetics of phylloquinone (vitamin K1) were evaluated in healthy human adult volunteers (15 male and 15 female) following oral and intravenous administration of a mixed micelles formulation (Konakion MM 2 mg) in an open label study design. The subjects were allocated to one of three genotype-specific groups (n = 10 in each group) in terms of VKORC1 promoter polymorphism c.-1639 G > A to explore the relationship between genotype and pharmacokinetic parameters. Blood samples were collected for up to 24 h after administration. Phylloquinone serum levels were determined by reversed phase HPLC with fluorometric detection after post-column zinc reduction. Pharmacokinetic evaluation was performed using non-compartmental analysis. Pharmacokinetic analysis of serum phylloquinone concentration versus time profiles revealed significant differences in the main pharmacokinetic parameters between groups. Upon oral administration, VKORC1 AG carriers showed 41 % higher mean bioavailability (p = 0.01) compared with homozygous AA individuals. Furthermore, AG subjects exhibited 30 % (p = 0.042) and 36 % (p = 0.021) higher mean AUC compared with GG and AA respectively. Terminal half-life was 32 % and 27 % longer for AG carriers in comparison to GG (p = 0.004) and AA (p = 0.015) genotypes respectively. Pharmacokinetic differences indicated significant inter-individual variance of vitamin K fate in the human body. The influence of the VKORC1 promoter polymorphism c.-1639 G > A on the pharmacokinetic properties of phylloquinone could be demonstrated in humans. To gain deeper insight in other potential genetic determinants of systemic vitamin K exposure, further correlation of the phenotype-genotype relationship of different players in vitamin K turnover has to be gained.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Sigma-Aldrich
Vitamin K1, viscous liquid
Supelco
Phytonadione, Pharmaceutical Secondary Standard; Certified Reference Material
Phytomenadione, European Pharmacopoeia (EP) Reference Standard
Supelco
Phylloquinone (K1), analytical standard