Merck
  • Comparative effects of in ovo exposure to sodium perchlorate on development, growth, metabolism, and thyroid function in the common snapping turtle (Chelydra serpentina) and red-eared slider (Trachemys scripta elegans).
Comparative effects of in ovo exposure to sodium perchlorate on development, growth, metabolism, and thyroid function in the common snapping turtle (Chelydra serpentina) and red-eared slider (Trachemys scripta elegans).
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2012-08-09)
Karen M Eisenreich, Karen M Dean, Mary Ann Ottinger, Christopher L Rowe
ABSTRAKT
Perchlorate is a surface and groundwater contaminant found in areas associated with munitions and rocket manufacturing and use. It is a thyroid-inhibiting compound, preventing uptake of iodide by the thyroid gland, ultimately reducing thyroid hormone production. As thyroid hormones influence metabolism, growth, and development, perchlorate exposure during the embryonic period may impact embryonic traits that ultimately influence hatchling performance. We topically exposed eggs of red-eared sliders (Trachemys scripta) and snapping turtles (Chelydra serpentina) to 200 and 177 μg/g of perchlorate (as NaClO(4)), respectively, to determine impacts on glandular thyroxine concentrations, embryonic growth and development, and metabolic rates of hatchlings for a period of 2 months post-hatching. In red-eared sliders, in ovo perchlorate exposure delayed hatching, increased external yolk size at hatching, increased hatchling mortality, and reduced total glandular thyroxine concentrations in hatchlings. In snapping turtles, hatching success and standard metabolic rates were reduced, liver and thyroid sizes were increased, and total glandular thyroxine concentrations in hatchlings were reduced after exposure to perchlorate. While both species were negatively affected by exposure, impacts on red-eared sliders were most severe, suggesting that the slider may be a more sensitive sentinel species for studying effects of perchlorate exposure to turtles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium perchlorate, ACS reagent, ≥98.0%