Merck
  • Enantioselective allylation of imines catalyzed by newly developed (-)-β-pinene-based π-allylpalladium catalyst: an efficient synthesis of (R)-α-propylpiperonylamine and (R)-pipecolic acid.
Enantioselective allylation of imines catalyzed by newly developed (-)-β-pinene-based π-allylpalladium catalyst: an efficient synthesis of (R)-α-propylpiperonylamine and (R)-pipecolic acid.
Organic & biomolecular chemistry (2012-08-23)
Rodney A Fernandes, Jothi L Nallasivam
ABSTRAKT
A newly developed π-allylpalladium with a (-)-β-pinene framework and an isobutyl side chain catalyzed the enantioselective allylation of imines in good yields and enantioselectivities (20 examples, up to 98% ee). An efficient enantioselective synthesis of the (R)-α-propyl piperonylamine part of DMP 777, a human leukocyte elastase inhibitor and (R)-pipecolic acid have been achieved as a useful application of this methodology.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Pipecolic acid, 99% (titration)
Sigma-Aldrich
Pipecolinic acid, 98%