Merck
Complete heart block associated with regadenoson: a real side effect.
Journal of nuclear cardiology : official publication of the American Society of Nuclear Cardiology (2012-08-24)
Anil Pandit, Samuel Unzek Freiman
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Aminophylline, ≥98%, powder