Merck
Sensitive fluorescence detection of etimicin based on derivatives of formaldehyde and acetylacetone.
Journal of fluorescence (2012-08-25)
Yafeng Zhuang, Lu Cao
ABSTRAKT
A novel fluorescence method for the determination of etimicin is described. Etimicin reacts with acetylacetone and formaldehyde in pH 4.0 Britton-Robinson (B.R.) buffer solution to from a fluorescent substance [I]. Emission spectra of [I] and the reagent blank were overlapped, so the arithmetic emission spectra of the fluorescent substance were obtained by subtracted form the spectra of [I] to the spectra of the reagent blank using the Fluorescence Data Software. There is a linear relationship between the intensity of the arithmetic emission spectra and the concentration of etimicin. Effects of pH, amount of acetylacetone-formaldehyde, and heating time on the determination of etimicin have been examined. Etimicin can be determined over the concentration range of 1.0 to 10.0 μg mL(-1) with a correlation coefficient of 0.9991. The relative standard deviation (RSD) for 11 repetitive determinations of 5.0 μg mL(-1) etimicin is 0.22%. The utility of this method was demonstrated by determining etimicin in commercial samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetylacetone, Wacker Chemie AG, ≥99.5% (GC)
Supelco
Acetylacetone, analytical standard
Sigma-Aldrich
Acetylacetone, ReagentPlus®, ≥99%