Merck
Characterization of vitellogenin and its derived yolk proteins in cloudy catshark (Scyliorhinus torazame).
Fish physiology and biochemistry (2012-08-25)
Kodai Yamane, Tomoki Yagai, Osamu Nishimiya, Rieko Sugawara, Haruna Amano, Toshiaki Fujita, Naoshi Hiramatsu, Takashi Todo, Takahiro Matsubara, Akihiko Hara
ABSTRAKT
Elasmobranchs (sharks and rays) exhibit unique reproductive characteristics and, in contrast to the situation in teleosts, very little is known about the identity, structure and physical characteristics of their egg yolk proteins. The aims of this study were to (1) detect and purify the vitellogenin (Vtg; egg yolk precursor) and yolk proteins (YPs) of the cloudy catshark (Scyliorhinus torazame), (2) examine the relationships between Vtg and YPs and (3) characterize and classify the deduced primary structure of the Vtg transcript (vtg). The apparent molecular weights of purified Vtg and putative Vtg-related YPs (lipovitellin: Lv, phosvitin: Pv) were determined by gel filtration and were ~560, >669 and ~58 kDa, respectively. Following SDS-PAGE, these purified products (i.e., Vtg, Lv and Pv) appeared as bands of ~210, ~110 and ~22 kDa, respectively. On Western blots, antisera against purified Vtg, Lv and Pv recognized the ~210 kDa Vtg band. Catshark Pv, in contrast to teleost Pvs, had a very low serine content. The catshark Vtg cDNA sequence (vtg) appeared to contain an open-reading frame consisting of domains encoding Lv, Pv and β'-component (β'-c). A phylogenetic analysis, with a consideration of genome duplication events, placed catshark vtg into the 'vtgAB type.' It is concluded that at least a single major type of Vtg protein, which is transcribed and translated from catshark vtgAB gene, is the precursor of three egg yolk proteins (Lv, Pv and β'-c) in catshark.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phosvitin from egg yolk