Merck
  • Isothiocyanates ameliorate the symptom of heart dysfunction and mortality in a murine AIDS model by inhibiting apoptosis in the left ventricle.
Isothiocyanates ameliorate the symptom of heart dysfunction and mortality in a murine AIDS model by inhibiting apoptosis in the left ventricle.
Journal of medicinal food (2012-08-29)
Jin-Nyoung Ho, Ho-Geun Yoon, Chang-Soo Park, Sunoh Kim, Woojin Jun, Ryowon Choue, Jeongmin Lee
ABSTRAKT
Cardiac involvement has been reported in as many as 45-55% of patients with human immunodeficiency virus (HIV) infection and acquired immune deficiency syndrome (AIDS), and significant cardiac morbidity is reported in 6-7% of HIV patients. We investigated the inhibitory effects of isothiocyanates (ITCs) on heart dysfunction and mortality by regulating apoptosis in the left ventricle of the heart in a murine AIDS model. Mice were divided into six groups: an uninfected group, an untreated LP-BM5 retrovirus-infected group, and four LP-BM5 retrovirus-infected groups treated with one of four ITCs (sulforaphane [SUL], indolo[3,2-b]carbazole, benzyl isothiocyanate [BITC], or phenethyl isothiocyanate [PEITC]). After 16 weeks, the median survival time of the LP-BM5 retrovirus-infected mice was 87 days, whereas that of the uninfected control group and all ITC treatment groups was over 112 days. SUL, PEITC, and BITC significantly inhibited apoptosis in the left ventricle by increasing the Bcl-2/Bax ratio compared with LP-BM5-infected mice. In addition, SUL and PEITC suppressed inducible nitric oxide synthase (iNOS) expression at both the mRNA and protein levels in the left ventricle of heart tissue infected with the LP-BM5 retrovirus by inactivating cytoplasmic nuclear factor κB (NF-κB). In conclusion, LP-BM5 retrovirus infection was related to survival of murine AIDS mice, and NF-κB-mediated iNOS expression may be an important mediator of left ventricle dysfunction of the heart. Furthermore, certain ITCs may have the potential to improve AIDS-related heart dysfunction due to their inhibition of apoptosis by decreasing iNOS and Bax expression through suppression of NF-κB.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Phenylethyl isothiocyanate, FG
Sigma-Aldrich
Phenethyl isothiocyanate, 99%