Merck
Spectroscopic investigations on Pr³+ and Nd³+ doped strontium-lithium-bismuth borate glasses.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-08-29)
D Rajesh, A Balakrishna, M Seshadri, Y C Ratnakaram
ABSTRAKT
Spectroscopic investigations on different concentrations (0.1, 0.5, 1.0, 1.5 and 2.0mol%) of Pr(3+) and Nd(3+) doped strontium lithium bismuth borate glasses have been done. X-ray diffraction, SEM with EDS, absorption and luminescence spectra were recorded for all the glass matrices and analyzed. X-ray diffraction profiles and SEM images conformed amorphous nature of investigated glass samples. EDS spectra of host glass and Pr(3+)doped glass matrices gave information about the chemical composition of glass samples. From the absorption spectra of Pr(3+) and Nd(3+) ions, Judd-Ofelt (J-O) intensity parameters (Ω(λ),λ=2, 4 and 6) have been calculated and compared with other glass matrices. The emission characteristics such as radiative lifetimes (τ(R)), measured and calculated branching ratios (β) and stimulated emission cross-sections (σ(P)) have been obtained for the observed emission transitions of Pr(3+) and Nd(3+) ions in the above glass matrix for all the concentrations. From the emission spectra of Pr(3+) and Nd(3+) doped glass matrices, the effect of concentration on the quenching of intensity of (1)D(2)→(3)H(4) transition of Pr(3+) ion and (4)F(3/2)→(4)I(9/2), (4)I(11/2) and (4)I(13/2) transitions of Nd(3+) have been studied and discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lithium metaborate, 99.995% trace metals basis
Sigma-Aldrich
Lithium metaborate, 99.9% trace metals basis
Sigma-Aldrich
Lithium metaborate, ACS reagent, ≥98.0%
Sigma-Aldrich
Lithium tetraborate, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium tetraborate, ≥99.995% trace metals basis