Merck
Design and optimization of a chronotherapeutic dosage form for treatment of nocturnal acid breakthrough.
Current drug delivery (2012-09-01)
Vaibhav Agarwal, Mayank Bansal
ABSTRAKT
Present work focuses on the use of tamarind gum to develop a drug delivery system making combined use of floating and pulsatile principles, for the chrono-prevention of nocturnal acid breakthrough. The desired aim was achieved by fabricating a floating delivery system bearing time - lagged coating of Tamarindus indica seed polymer for the programmed release of Famotidine. Response Surface METHODology was the statistical tool that was employed for experiment designing, mathematical model generation and optimization study. A 32 full factorial design was used in designing the experiment. % weight ratio of tamarind gum to ethyl cellulose in the coating combination and the coating weight were the independent variables, whereas the lag time for drug release and the cumulative % drug release in 330 minutes were the observed responses. Results revealed that both the coating composition and the coating weight significantly affected the release of drug from the dosage form. The optimized formulation prepared according to the computer generated software, Design-Expert® deciphered response which were in close proximity with the experimental responses, thus confirming the robustness and accuracy of the predicted model for the utilization of natural polymer like tamarind gum for the chronotherapeutic treatment of nocturnal acid breakthrough.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Famotidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Famotidine