Merck
Identification and quantification of key microbial trophic groups of methanogenic glucose degradation in an anaerobic digester sludge.
Bioresource technology (2012-09-05)
Tsukasa Ito, Kazumi Yoshiguchi, Herto Dwi Ariesyady, Satoshi Okabe
ABSTRAKT
We investigated the major phylogenetic groups and population size of glucose-, propionate-, and acetate-degrading bacteria in the glucose-degrading anaerobic digester sludge by stable-isotope probing analysis of 16S rRNA (RNA-SIP) with [(13)C(6)]glucose followed by time course analysis of microautoradiography combined with fluorescent in situ hybridization (MAR-FISH) with [U-(14)C]glucose. The results indicated that glucose was predominately degraded to CH(4) and CO(2) by glucose-degrading Propionibacterium and Olsenella that are belonging to the phylum Actinobacteria, propionate-degrading Smithella and Syntrophobacter, and acetate-degrading Methanosaeta and Synergistes group 4 in this anaerobic sludge. The population size of propionate degraders was the smallest among three trophic groups and the specific degradation rate of propionate was also low. The specific degradation rate of acetate was low even though their population size was comparable to the glucose degraders. These results could explain why the degradation of propionate and acetate was the rate-limiting step in methanogenic glucose degradation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, ≥99.5%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~98%