Merck
Mitogen-activated protein kinase (MEK/ERK) inhibition sensitizes cancer cells to centromere-associated protein E inhibition.
International journal of cancer (2012-09-06)
Patrick A Mayes, Yan Y Degenhardt, Andrew Wood, Yana Toporovskya, Sharon J Diskin, Elizabeth Haglund, Christopher Moy, Richard Wooster, John M Maris
ABSTRAKT
Inhibition of centromere-associated protein-E (CENP-E) has demonstrated preclinical anti-tumor activity in a number of tumor types including neuroblastoma. A potent small molecule inhibitor of the kinesin motor activity of CENP-E has recently been developed (GSK923295). To identify an effective drug combination strategy for GSK923295 in neuroblastoma, we performed a screen of siRNAs targeting a prioritized set of genes that function in therapeutically tractable signaling pathways. We found that siRNAs targeted to extracellular signal-related kinase 1 (ERK1) significantly sensitized neuroblastoma cells to GSK923295-induced growth inhibition (p = 0.01). Inhibition of ERK1 activity using pharmacologic inhibitors of mitogen-activated ERK kinase (MEK1/2) showed significant synergistic growth inhibitory activity when combined with GSK923295 in neuroblastoma, lung, pancreatic and colon carcinoma cell lines. Synergistic growth inhibitory activity of combined MEK/ERK and CENP-E inhibition was a result of increased mitotic arrest and apoptosis. There was a significant correlation between ERK1/2 phosphorylation status in neuroblastoma cell lines and GSK923295 growth inhibitory activity (r = 0.823, p = 0.0006). Consistent with this result we found that lung cancer cell lines harboring RAS mutations, which leads to oncogenic activation of MEK/ERK signaling, were significantly more resistant than cell lines with wild-type RAS to GSK923295-induced growth inhibition (p = 0.047). Here we have identified (MEK/ERK) activity as a potential biomarker of relative GSK923295 sensitivity and have shown the synergistic effect of combinatorial MEK/ERK pathway and CENP-E inhibition across different cancer cell types including neuroblastoma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sarcosine, BioXtra
Sigma-Aldrich
Sarcosine, 98%
Sigma-Aldrich
Sarcosine, crystallized, ≥98.0% (T)