Merck
  • A versatile fluorescence approach to kinetic studies of hydrocarbon autoxidations and their inhibition by radical-trapping antioxidants.
A versatile fluorescence approach to kinetic studies of hydrocarbon autoxidations and their inhibition by radical-trapping antioxidants.
Chemical communications (Cambridge, England) (2012-09-07)
Jason J Hanthorn, Evan Haidasz, Paul Gebhardt, Derek A Pratt
ABSTRAKT
We report a simple coumarin-triarylphosphine conjugate that undergoes fluorescence enhancement upon reaction with hydroperoxides and demonstrate its use to follow autoxidations of 7-dehydrocholesterol (at 37 °C) and hexadecane (at 160 °C) and their inhibition by antioxidants.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
7-Dehydrocholesterol, ≥95.0% (HPLC)
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Supelco
Hexadecane, analytical standard