Merck
Neuroprotective effect of sinapic acid in a mouse model of amyloid β(1-42) protein-induced Alzheimer's disease.
Pharmacology, biochemistry, and behavior (2012-09-14)
Hyung Eun Lee, Dong Hyun Kim, Se Jin Park, Jong Min Kim, Young Woo Lee, Jun Man Jung, Chang Hwan Lee, Jin Gyu Hong, Xiaotong Liu, Mudan Cai, Keon Ju Park, Dae Sik Jang, Jong Hoon Ryu
ABSTRAKT
Sinapic acid (SA) is a phenylpropanoid compound with anti-inflammatory and neuroprotective activities. The neuroprotective effects of SA in a mouse model of amyloid β (Aβ)(1-42) protein-induced Alzheimer's disease (AD) were investigated. Mice received a bilateral injection of Aβ(1-42) protein into the hippocampus to verify the efficacy of SA. Mice were treated with SA (10mg/kg/day, p.o.) for 7days beginning immediately after Aβ(1-42) protein injection, and an acquisition trial of the passive avoidance task was conducted 1h after the last administration of SA. Retention trial was conducted 24h after the acquisition trial, and mice were sacrificed for immunohistochemistry immediately after the retention trial. SA rescued neuronal cell death in the hippocampal CA1 region and also attenuated the increase of iNOS expression, glial cell activations and nitrotyrosine expressions induced by Aβ(1-42) protein. SA significantly attenuated memory impairment in the passive avoidance task. These results suggest that SA ameliorated Aβ(1-42) protein-related pathology including neuronal cell death and cognitive dysfunction via its anti-oxidative and anti-inflammatory activities, and may be an efficacious treatment for AD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
Sinapic acid, ≥98%, powder