Merck
Study of the solvent effects on the molecular structure and CO stretching vibrations of flurbiprofen.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-09-19)
Nalan Tekin, Hacer Pir, Seda Sagdinc
ABSTRAKT
The effects of 15 solvents on the C=O stretching vibrational frequency of flurbiprofen (FBF) were determined to investigate solvent-solute interactions. Solvent effects on the geometry and C=O stretching vibrational frequency, ν(C=O), of FBF were studied theoretically at the DFT/B3LYP and HF level in combination with the polarizable continuum model and experimentally using attenuated total reflection infrared spectroscopy (ATR-IR). The calculated C=O stretching frequencies in the liquid phase are in agreement with experimental values. Moreover, the wavenumbers of ν(C=O) of FBF in different solvents have been obtained and correlated with the Kirkwood-Bauer-Magat equation (KBM), the solvent acceptor numbers (ANs), and the linear solvation energy relationships (LSERs). The solvent-induced stretching vibrational frequency shifts displayed a better correlation with the LSERs than with the ANs and KBM.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Flurbiprofen, cyclooxygenase inhibitor