Merck
Organic-inorganic hybrid mesoporous polymers fabricated by using (CTA)2S2O8 as self-decomposed soft templates.
Langmuir : the ACS journal of surfaces and colloids (2012-10-03)
Tianyou Chen, Binyang Du, Zhiqiang Fan
ABSTRAKT
Organic-inorganic hybrid mesoporous polymers were successfully synthesized by using a template-directed free radical polymerization technique in aqueous solution at 0-5 °C with oxidative complexes as self-decomposed soft templates. The oxidative complexes ((CTA)(2)S(2)O(8)), which were formed between anionic oxidant (S(2)O(8)(2-)) and cationic surfactant (cetyltrimethylammonium bromide, CTAB) at 0-5 °C, can be automatically decomposed due to the reduction of S(2)O(8)(2-). No additional treatment was needed to remove the templates. The reactive functional monomer, 3-(trimethoxysilyl)propyl methacrylate (TMSPMA), was used as main monomer. Styrene was used as the comonomer. With simultaneous free radical copolymerization of TMSPMA and styrene, condensation of methoxysilyl groups, and the self-decomposition of (CTA)(2)S(2)O(8), organic-inorganic hybrid mesoporous polymers were successfully obtained. The mesoporous structures and morphologies of the resultant hybrid mesoporous polymers were found to be strongly dependent on the feed amounts of TMSPMA and styrene. In the absence of styrene, the hybrid polymer PTMSPMA exhibited mesh-like bicontinuous structures with mesopores and high surface area (335 m(2)/g). With the incorporation of styrene, mesoporous nanoparticles were obtained. The surface areas of the mesoporous nanoparticles decreased with the increase of styrene contents. The adsorption capabilities of such mesoporous polymers for organic dye (Congo red) and protein (bovine serum albumin) were also studied.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Potassium persulfate, ACS reagent, ≥99.0%