Merck
  • Immobilization of unraveled immunoglobulin G using well-oriented ZZ-His protein on functionalized microtiter plate for sensitive immunoassay.
Immobilization of unraveled immunoglobulin G using well-oriented ZZ-His protein on functionalized microtiter plate for sensitive immunoassay.
Analytical biochemistry (2012-10-03)
Hong-Ming Yang, Shu-Juan Liang, Jin-Bao Tang, Yong Chen, Yuan-Zheng Cheng
ABSTRAKT
Highly efficient protein immobilization is extremely crucial for solid-phase immunoassays. We present a strategy for oriented immobilization of functionally intact immunoglobulin G (IgG) on a polystyrene microtiter plate via iminodiacetic acid (IDA)-Ni(2+) and ZZ-His protein interaction. We immobilized a ZZ-EAP (Escherichia coli alkaline phosphatase)-His fusion protein, which exhibits Fc binding, His tag, and intrinsic AP activities, and analyzed it against the interaction between rabbit IgG anti-horseradish peroxidase (anti-HRP) and its binding partner HRP to investigate the specificity and efficacy of this method. We compared the IDA-Ni(2+)-(ZZ-His) method with ZZ-EAP random immobilization using sandwich enzyme-linked immunosorbent assay, and the results showed that the former method had an enhanced signal, 10-fold higher sensitivity, and a wider linear range. Thus, the proposed method allows a broad range of oriented immobilized functionally intact IgG antibodies on polystyrene plates using only one type of IDA-Ni(2+) chelate surface because the ZZ protein can bind to the Fc region of various IgGs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Iminodiacetic acid, 98%
Sigma-Aldrich
Iminodiacetic acid, purum, ≥98.0% (T)