Merck
  • Characterization and comparison of methacrylic acid with 2-acrylamido-2-methyl-1-propanesulfonic acid in the preparation of monolithic column for capillary electrochromatography.
Characterization and comparison of methacrylic acid with 2-acrylamido-2-methyl-1-propanesulfonic acid in the preparation of monolithic column for capillary electrochromatography.
Journal of chromatographic science (2012-10-09)
Daisuke Horiguchi, Kaname Ohyama, Tomoko Masunaga, Yoshiko Fujita, Marwa Fathy Bakr Ali, Naoya Kishikawa, Naotaka Kuroda
ABSTRAKT
Butyl methacrylate (BMA)-ethylene dimethacrylate (EDMA)-methacrylic acid (MAA) and BMA-EDMA-2-acrylamido-2-methyl-1-propanesulfonic acid (AMPS) monolithic columns were prepared by varying the percentage of ionic monomers for capillary electrochromatography. Monolithic columns with a higher content of ionic monomers provided better column efficiency, and the performance of BMA-EDMA-MAA monoliths was better than BMA-EDMA-AMPS. To characterize and optimize BMA-EDMA-MAA monoliths, the effects of the content of cross-linker and the total monomer in the polymerization mixture on column performance were also studied. Plate heights of 8.2 µm for the unretained solute (thiourea) and 12.6 µm for the retained solute (naphthalene) were achieved with a monolithic column using 2.5% MAA (Column I).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Acrylamido-2-methyl-1-propanesulfonic acid sodium salt solution, 50 wt. % in H2O
Sigma-Aldrich
2-Acrylamido-2-methyl-1-propanesulfonic acid, 99%
Sigma-Aldrich
Sodium methacrylate, 99%
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%