Merck
  • Ethylenediammonium dication: H-bonded complexes with terephthalate, chloroacetate, phosphite, selenite and sulfamate anions. Detailed vibrational spectroscopic and theoretical studies of ethylenediammonium terephthalate.
Ethylenediammonium dication: H-bonded complexes with terephthalate, chloroacetate, phosphite, selenite and sulfamate anions. Detailed vibrational spectroscopic and theoretical studies of ethylenediammonium terephthalate.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-10-20)
M K Marchewka, M Drozd
ABSTRAKT
Crystalline complexes between ethylenediammonium dication and terephthalate, chloroacetate, phosphite, selenite and sulfamate anions were obtained by slow evaporation from water solution method. Room temperature powder infrared and Raman measurements were carried out. For ethylenediammonium terephthalate theoretical calculations of structure were performed by two ways: ab-initio HF and semiempirical PM3. In this case the PM3 method gave more accurate structure (closer to X-ray results). The additional PM3 calculations of vibrational spectra were performed. On the basis theoretical approach and earlier vibrational studies of similar compounds the vibrational assignments for observed bands have been proposed. All compounds were checked for second harmonic generation (SHG).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Chloroacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Terephthalic acid, 98%
Sigma-Aldrich
Chloroacetic acid, 99%
Sigma-Aldrich
Chloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfamate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium sulfamate, BioXtra, ≥98.0%
Sigma-Aldrich
Sulfamic acid, ACS reagent, 99.3%
Supelco
Sulfamic acid, analytical standard (for acidimetry), ACS reagent
Sigma-Aldrich
Sulfamic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulfamic acid, reagent grade, 98%
Sigma-Aldrich
Sulfamic acid, 99.999% trace metals basis