Merck
  • The exposure of highly toxic aconitine does not significantly impact the activity and expression of cytochrome P450 3A in rats determined by a novel ultra performance liquid chromatography-tandem mass spectrometric method of a specific probe buspirone.
The exposure of highly toxic aconitine does not significantly impact the activity and expression of cytochrome P450 3A in rats determined by a novel ultra performance liquid chromatography-tandem mass spectrometric method of a specific probe buspirone.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-10-23)
Lijun Zhu, Xiaoshan Yang, Juan Zhou, Lan Tang, Bijun Xia, Ming Hu, Fuyuan Zhou, Zhongqiu Liu
ABSTRAKT
Aconitum species are widely used to treat rheumatism, cardiovascular diseases, and tumors in China and other Asian countries. The herbs are always used with drugs such as paclitaxel. Aconitine (AC) is one of the main bioactive/high-toxic alkaloids of Aconitum roots. AC is metabolized by cytochrome P450 (CYP) 3A. However, whether AC inhibits/induces CYP3A, which causes drug-drug interaction (DDI) is unclear. Our study aims to explore the potent effects of AC, as a marker component of Aconitum, on CYP3A using the probe buspirone in rats. The effects of oral AC on pharmacokinetics of buspirone were evaluated. CYP3A activity and protein levels in rat liver microsomes pretreated with oral AC were also measured using in vitro buspirone metabolism and Western blot. Buspirone and its major metabolites 1-(2-pyrimidinyl)piperazine and 6'-hydroxybuspirone were determined using a newly validated UPLC-MS/MS method. Single dose and 7-day AC administration at 0.125mg/kg had no effect on CYP3A activity since no change in the formation of 1-(2-pyrimidinyl)piperazine and 6'-hydroxybuspirone. CYP3A activity and protein levels in liver microsomes were also not affected by 7-day AC pretreatment at 0.125mg/kg. Therefore, AC neither inhibits nor induces CYP3A in rats, indicating AC does not cause CYP3A-related DDI in the liver.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Buspirone hydrochloride
Buspirone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aconitine, ≥95% (HPLC), crystalline