- A furan Diels-Alder cycloaddition approach to scyphostatin analogues.
A furan Diels-Alder cycloaddition approach to scyphostatin analogues.
Organic & biomolecular chemistry (2012-10-23)
Calum J Fraser, Gareth P Howell, Joseph P A Harrity
PMID23086598
ABSTRAKT
The synthesis of two diastereoisomers of the epoxycyclohexenone core of scyphostatin, a naturally occurring sphingomyelinase inhibitor, has been achieved via a common oxabicyclic intermediate. The diastereomeric intermediates are accessed by stereodivergent oxidative functionalisation processes, followed by a Lewis acid mediated ring opening rearrangement reaction.