Merck
The clinical significance of NOTCH1 and SF3B1 mutations in the UK LRF CLL4 trial.
Blood (2012-10-23)
David G Oscier, Matthew J J Rose-Zerilli, Nils Winkelmann, David Gonzalez de Castro, Belen Gomez, Jade Forster, Helen Parker, Anton Parker, Anne Gardiner, Andrew Collins, Monica Else, Nicholas C P Cross, Daniel Catovsky, Jonathan C Strefford
ABSTRAKT
NOTCH1 and SF3B1 mutations have been previously reported to have prognostic significance in chronic lymphocytic leukemia but to date they have not been validated in a prospective, controlled clinical trial. We have assessed the impact of these mutations in a cohort of 494 patients treated within the randomized phase 3 United Kingdom Leukaemia Research Fund Chronic Lymphocytic Leukemia 4 (UK LRF CCL4) trial that compared chlorambucil and fludarabine with and without cyclophosphamide in previously untreated patients. We investigated the relationship of mutations in NOTCH1 (exon 34) and SF3B1 (exon 14-16) to treatment response, survival and a panel of established biologic variables. NOTCH1 and SF3B1 mutations were found in 10% and17% of patients, respectively. NOTCH1 mutations correlated with unmutated IGHV genes, trisomy 12, high CD38/ ZAP-70 expression and were associated with reduced overall (median 54.8 vs 74.6 months, P = .02) and progression-free (median 22.0 vs 26.4 months, P = .02) survival. SF3B1 mutations were significantly associated with high CD38 expression and with shorter overall survival (median 54.3 vs 79.0 months, P < .001). Furthermore, multivariate analysis, including baseline clinical variables, treatment, and adverse prognostic factors demonstrated that although TP53 alterations remained the most informative marker of dismal survival in this cohort, NOTCH1 (HR 1.58, P = .03) and SF3B1 (HR 1.52, P = .01) mutations have added independent prognostic value.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chlorambucil