- Moth-eaten appearance of tubulointerstitial nephritis and uveitis syndrome on (99m)technetium dimercaptosuccinic acid scintigraphy.
Moth-eaten appearance of tubulointerstitial nephritis and uveitis syndrome on (99m)technetium dimercaptosuccinic acid scintigraphy.
The Journal of pediatrics (2012-10-25)
Masaki Shimizu, Sayaka Ishikawa, Kazuyuki Ueno, Akihiro Yachie
PMID23092525
MATERIAŁY