Merck
  • Efficacy of budesonide in combination with formoterol in patients with inadequately controlled asthma on fluticasone in combination with salmeterol.
Efficacy of budesonide in combination with formoterol in patients with inadequately controlled asthma on fluticasone in combination with salmeterol.
Allergology international : official journal of the Japanese Society of Allergology (2012-10-25)
Yoshihiro Kanemitsu, Hideo Kita, Akio Niimi, Yoshinori Fuseya, Kazuya Tanimura, Yuko Katayama, Tamaki Takahashi, Yukimasa Hatachi, Yumi Nishihara, Toshikatsu Sado
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Salmeterol xinafoate, ≥98% (HPLC), solid