Merck
Mechanisms of peptide-induced pore formation in lipid bilayers investigated by oriented 31P solid-state NMR spectroscopy.
PloS one (2012-10-25)
Kresten Bertelsen, Jerzy Dorosz, Sara Krogh Hansen, Niels Chr Nielsen, Thomas Vosegaard
ABSTRAKT
There is a considerable interest in understanding the function of antimicrobial peptides (AMPs), but the details of their mode of action is not fully understood. This motivates extensive efforts in determining structural and mechanistic parameters for AMP's interaction with lipid membranes. In this study we show that oriented-sample (31)P solid-state NMR spectroscopy can be used to probe the membrane perturbations and disruption by AMPs. For two AMPs, alamethicin and novicidin, we observe that the majority of the lipids remain in a planar bilayer conformation but that a number of lipids are involved in the peptide anchoring. These lipids display reduced dynamics. Our study supports previous studies showing that alamethicin adopts a transmembrane arrangement without significant disturbance of the surrounding lipids, while novicidin forms toroidal pores at high concentrations leading to more extensive membrane disturbance.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Alamethicin from Trichoderma viride, ≥98% (HPLC)
Sigma-Aldrich
Alamethicin, Ready Made Solution from Trichoderma viride, 5 mg/mL in DMSO