Merck
  • Expression of the calcium sensing receptor in human peripheral blood T lymphocyte and its contribution to cytokine secretion through MAPKs or NF-κB pathways.
Expression of the calcium sensing receptor in human peripheral blood T lymphocyte and its contribution to cytokine secretion through MAPKs or NF-κB pathways.
Molecular immunology (2012-10-30)
Tingting Li, Mingrui Sun, Xin Yin, Chunli Wu, Qiuyue Wu, Shanli Feng, Hong Li, Ying Luan, Jie Wen, Lixin Yan, Binhui Zhao, Changqing Xu, Yihua Sun
ABSTRAKT
The calcium-sensing receptor (CaSR) has been reported to play an important role in many tissues and organs. However, studies about the expression and function of CaSR in T lymphocytes are still not very lucid. In this study, we investigated the above-mentioned issues using RT-PCR, immunofluorescence staining, Western blotting, and the ELISA techniques. We found that the CaSR protein was expressed, and mainly located in the membrane in the normal human peripheral blood T lymphocytes. GdCl(3) (an agonist of CaSR) increased the dose-dependency of the CaSR expression, which was abolished by NPS2390 (an inhibitor of CaSR). GdCl(3) and Ca(2+) increased the phosphorylation of extracellular signal-regulated kinase (ERK)1/2 (one subgroup of MAPKs) and P65 (subunit of NF-κB),but, they had no significant effects on the JNK and P38 subgroups of MAPKs. Meantime, GdCl(3) and Ca(2+) stimulated both the IL-6 and TNF-β releases and their mRNA expressions. However, these effects of GdCl(3) and Ca(2+) were inhibited by NPS2390, U0126 (MAPKs pathway inhibitor) or Bay-11-7082 (NF-κB pathway inhibitor). These results suggested that CaSR was functionally expressed in the T cells, and the activated CaSR contributed to the cytokine secretion through the partial MAPK and NF-κB pathways.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Gadolinium(III) chloride, powder, 99.99% trace metals basis