Merck
Anti-Candida albicans effectiveness of citral and investigation of mode of action.
Pharmaceutical biology (2012-11-03)
Igara Oliveira Lima, Fernanda de Medeiros Nóbrega, Wylly Araújo de Oliveira, Edeltrudes de Oliveira Lima, Everardo Albuquerque Menezes, Francisco Afrânio Cunha, Margareth de Fátima Formiga Melo Diniz
ABSTRAKT
Candidiasis is a mycosis caused by Candida species, which is of clinical importance due to the increase in resistant yeasts. Candida infection has been a serious health problem due to the inappropriate use of antibiotics. Therefore, it is necessary to study molecules with an antifungal action. Citral is a monoterpene with known pharmacological properties, including antimicrobial action. The aim of this work was to determine the minimum inhibitory concentration (MIC) and minimum fungicidal concentration (MFC) of citral and the probable mode of action. The MIC of citral was determined by the broth microdilution method using Sabouraud dextrose medium. Additionally, the interference of citral in cell wall (sorbitol assay) and the binding of citral to ergosterol and cholesterol were studied, carried out by broth microdilution method. The MIC and MFC of citral were 512 and 1024 µg/mL, respectively. The MIC of amphotericin B was 1 µg/mL. The mechanism of action did not involve either the cell wall or ergosterol. However, the presence of cholesterol increased the MIC of citral to 1024 µg/mL, indicating there is some interaction between citral and cholesterol. Amphotericin B was used as the positive control, and it showed a high MIC in the presence of ergosterol (32 µg/mL), while in the presence of cholesterol MIC increased to 4 µg/mL. Citral inhibits the growth of C. albicans. The probable mechanism of action did not involve the cell wall or ergosterol. Citral is able to interact with cholesterol. More studies are necessary to describe their effects completely.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Citral, natural, ≥96%, FCC, FG
Sigma-Aldrich
Citral, 95%
Sigma-Aldrich
Citral, mixture of cis and trans, ≥96%, FG