Merck
Peptide-induced bilayer thinning structure of unilamellar vesicles and the related binding behavior as revealed by X-ray scattering.
Biochimica et biophysica acta (2012-11-06)
Chun-Jen Su, Shiuan-Shiaou Wu, U-Ser Jeng, Ming-Tao Lee, An-Chung Su, Kuei-Fen Liao, Wei-Yu Lin, Yu-Shan Huang, Chun-Yu Chen
ABSTRAKT
We have studied the bilayer thinning structure of unilamellar vesicles (ULV) of a phospholipid 1,2-dierucoyl-sn-glycero-3-phosphocholine (di22:1PC) upon binding of melittin, a water-soluble amphipathic peptide. Successive thinning of the ULV bilayers with increasing peptide concentration was monitored via small-angle X-ray scattering (SAXS). Results suggest that the two leaflets of the ULV of closed bilayers are perturbed and thinned asymmetrically upon free peptide binding, in contrast to the centro-symmetric bilayer thinning of the substrate-oriented multilamellar membranes (MLM) with premixed melittin. Moreover, thinning of the melittin-ULV bilayer associates closely with peptide concentration in solution and saturates at ~4%, compared to the ~8% maximum thinning observed for the correspondingly premixed peptide-MLM bilayers. Linearly scaling the thinning of peptide-ULV bilayers to that of the corresponding peptide-MLM of a calibrated peptide-to-lipid ratio, we have deduced the number of bound peptides on the ULV bilayers as a function of free peptide concentration in solution. The hence derived X-ray-based binding isotherm allows extraction of a low binding constant of melittin to the ULV bilayers, on the basis of surface partition equilibrium and the Gouy-Chapman theory. Moreover, we show that the ULV and MLM bilayers of di22:1PC share a same thinning constant upon binding of a hydrophobic peptide alamethicin; this result supports the linear scaling approach used in the melittin-ULV bilayer thinning for thermodynamic binding parameters of water-soluble peptides.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Alamethicin, Ready Made Solution from Trichoderma viride, 5 mg/mL in DMSO
Sigma-Aldrich
Melittin, ≥97% (HPLC), synthetic
Sigma-Aldrich
Alamethicin from Trichoderma viride, ≥98% (HPLC)
Sigma-Aldrich
Melittin from honey bee venom, ≥85% (HPLC)
Sigma-Aldrich
Melittin from honey bee venom, ≥65% (HPLC)