Merck
Butyrolactone and cycloheptanetrione from mangrove-associated fungus Aspergillus terreus.
Chemical & pharmaceutical bulletin (2012-11-06)
Yi Shen, Jianhua Zou, Dan Xie, Hanlin Ge, Xiuping Cao, Jungui Dai
ABSTRAKT
A new butyrolactone, 7″-hydroxybutyrolactone III (1) and three new cycloheptanetriones, terretrione A-C (2-4), together with five known compounds, butyrolactone I, cyclo(Leu-Pro), cyclo(Val-Pro), cyclo(Ile-Pro), cyclo(Phe-Pro), were isolated from mangrove-associated marine fungus Aspergillus terreus. The structures of these compounds were elucidated on the basis of physical data analysis (NMR, high resolution-electrospray ionization (HR-ESI)-MS), especially by 2D-NMR techniques. These compounds showed weak cytotoxicity in vitro against HCT-8, Bel-7402, BGC-823, A2780 cell lines.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tropolone, 98%