Merck
  • Molecular analysis of the rare in(Lu) blood type: toward decoding the phenotypic outcome of haploinsufficiency for the transcription factor KLF1.
Molecular analysis of the rare in(Lu) blood type: toward decoding the phenotypic outcome of haploinsufficiency for the transcription factor KLF1.
Human mutation (2012-11-06)
Virginie Helias, Carole Saison, Thierry Peyrard, Eliane Vera, Claude Prehu, Jean-Pierre Cartron, Lionel Arnaud
ABSTRAKT
KLF1 encodes an erythroid transcription factor, whose essential function in erythropoiesis has been demonstrated by extensive studies in mouse models. The first reported mutations in human KLF1 were found in individuals with a rare and asymptomatic blood type called In(Lu). Here, we show that KLF1 haploinsufficiency is responsible for the In(Lu) blood type, after redefining this peculiar blood type using flow cytometry to quantify the levels of BCAM and CD44 on red blood cells. We found 10 (seven novel) heterozygous KLF1 mutations responsible for the In(Lu) blood type. Although most were obligate loss-of-function mutations due to the truncation of the DNA-binding domain of KLF1, three were missense mutations that were located in its DNA-binding domain and impaired the transactivation capacity of KLF1 in vitro. We further showed that the levels of the hemoglobin variants HbF and HbA(2) were increased in the In(Lu) blood type, albeit differently. The levels of the membrane glycoproteins BCAM and CD44 were also differently reduced on In(Lu) red blood cells. This biochemical and genetic analysis of the In(Lu) blood type tackles the phenotypic outcome of haploinsufficiency for a transcription factor.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hemoglobin A2, Ferrous Stabilized human, lyophilized powder
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse