Merck
Non-specific inhibitors of aquaporin-4 stimulate S100B secretion in acute hippocampal slices of rats.
Brain research (2012-11-13)
Caroline Zanotto, Renata Torres Abib, Cristiane Batassini, Lucas Silva Tortorelli, Regina Biasibetti, Letícia Rodrigues, Patrícia Nardin, Fernanda Hansen, Carmem Gottfried, Marina Concli Leite, Carlos-Alberto Gonçalves
ABSTRAKT
Aquaporin-4 (AQP-4) is the principal brain water channel and is predominantly expressed in astrocytes suggesting its dynamic involvement in water homeostasis in brain tissue. Due to the co-localization of AQP-4 and inward rectifier K(+) channels Kir 4.1, a functional coupling between these proteins has been proposed. AQP-4 has a putative role in the physiopathology of brain disorders including epilepsy and trauma. S100B is a calcium-binding protein expressed and secreted by astrocytes, and commonly used as a parameter of astroglial activation. Here, we investigate a possible link between AQP-4 activity (and Kir 4.1) and S100B secretion in hippocampal slices of rats of different ages using non-specific inhibitors of AQP-4 (AZA, acetazolamide and TEA, tetraethylammonium) and Kir 4.1 (barium chloride). We found that blockade of AQP-4 with TEA and AZA produced an increase in S100B secretion in young rats, compatible with an astroglial activation observed in many conditions of brain injury. On the other hand, BaCl(2) induced Kir 4.1 inhibition caused a decrease in S100B secretion. Both channels, AQP-4 and Kir 4.1, exhibited a similar ontogenetic profile, in spite of the functional uncoupling, in relation to S100B secretion. Moreover, we found a significant increase in the S100B secretion basal levels with the increasing of animal age and the incubation with high levels of potassium resulted in a decrease of S100B secretion in 30 and 90-day old rats. These data, together with previous observations from gap junctions and glutamate transport of astrocytes, contribute to characterize the operational system involving astroglial activation, particularly on S100B secretion, in brain disorders.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Acetazolamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetazolamide, ≥99%, powder
Sigma-Aldrich
Barium chloride dihydrate, ≥99%
Sigma-Aldrich
Barium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Barium chloride dihydrate, ≥99.999% trace metals basis
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.95% trace metals basis
Sigma-Aldrich
Barium chloride, 99.9% trace metals basis
Sigma-Aldrich
Barium chloride, 99.999% trace metals basis
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis