Merck
  • Simultaneous determination of the combined drugs of ceftriaxone sodium, metronidazole, and levofloxacin in human urine by high-performance liquid chromatography.
Simultaneous determination of the combined drugs of ceftriaxone sodium, metronidazole, and levofloxacin in human urine by high-performance liquid chromatography.
Journal of clinical laboratory analysis (2012-11-13)
Hanwen Sun, Hongliu Wang, Xusheng Ge
ABSTRAKT
To develop a new high-performance liquid chromatography (HPLC) method for simultaneous determination of the combined drugs (ceftriaxone sodium, metronidazole, and levofloxacin) in human urine. Ceftriaxone sodium, metronidazole, and levofloxacin were separated on a Kromasil 100-5 C18 (250 mm × 4.6 mm, 5 μm, AKZO NOBEL, Bohus, Sweden) analytical column, using the mobile phase consisted of 1.5 mM KH(2) PO(4) (pH 4.5) with 0.0125% triethylamine-methnol (70:30, v/v). Ceftriaxone sodium, metronidazole, and levofloxacin were detected by a photodiode-array detector at 247, 320, 292 nm, respectively. Under optimal conditions, the effective separation of ceftriaxone sodium, metronidazole, and levofloxacin was achieved. A good linearity with the correlation coefficients more than 0.999 was demonstrated. The detection limits of ceftriaxone sodium, metronidazole, and levofloxacin were 0.05, 0.01, and 0.25 μg/ml, respectively, and the average recoveries in human urine were in the range from 97.73 to 100.7% with the average relative standard deviation (RSD) in the range of 2.5% and 3.0%. The proposed method was sensitive, accurate, and rapid. This work may provide a reference for clinical rational drug use and methodology for the pharmacokinetics study of the combined drugs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium Phosphate Monobasic solution, reagent grade, 1.0 M in solution
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%