Merck
[Pneumonia due to Yersinia enterocolitica].
Medecine et maladies infectieuses (2012-12-15)
P-A Jarrot, R Mahmoudi, J-L Novella, P Manckoundia
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cefotaxime sodium salt, suitable for plant cell culture, BioReagent, powder or crystals
Sigma-Aldrich
Cefotaxime sodium salt, potency: 916-964 μg per mg
Cefotaxime sodium, European Pharmacopoeia (EP) Reference Standard