Merck
Catalytic conversion of furfural into a 2,5-furandicarboxylic acid-based polyester with total carbon utilization.
ChemSusChem (2012-12-15)
Tao Pan, Jin Deng, Qing Xu, Yong Zuo, Qing-Xiang Guo, Yao Fu
ABSTRAKT
One divided into two combined into one: The catalytic conversion of furfural into a 2,5-furandicarboxylic acid-based polyester, linked by the disproportionation of furoate to furan and 2,5-furandicarboxylate, is reported. In this manner, all carbons are utilized, demonstrating the success of combining a platform molecule from C(5) sugars (furfural) to one from C(6) sugars (2,5-FDCA).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Furfural, ≥98%, FCC, FG