Merck
Translation elongation factor EF-P alleviates ribosome stalling at polyproline stretches.
Science (New York, N.Y.) (2012-12-15)
Susanne Ude, Jürgen Lassak, Agata L Starosta, Tobias Kraxenberger, Daniel N Wilson, Kirsten Jung
ABSTRAKT
Translation elongation factor P (EF-P) is critical for virulence in bacteria. EF-P is present in all bacteria and orthologous to archaeal and eukaryotic initiation factor 5A, yet the biological function has so far remained enigmatic. Here, we demonstrate that EF-P is an elongation factor that enhances translation of polyproline-containing proteins: In the absence of EF-P, ribosomes stall at polyproline stretches, whereas the presence of EF-P alleviates the translational stalling. Moreover, we demonstrate the physiological relevance of EF-P to fine-tune the expression of the polyproline-containing pH receptor CadC to levels necessary for an appropriate stress response. Bacterial, archaeal, and eukaryotic cells have hundreds to thousands of polyproline-containing proteins of diverse function, suggesting that EF-P and a/eIF-5A are critical for copy-number adjustment of multiple pathways across all kingdoms of life.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Poly-L-proline, mol wt >30,000
Sigma-Aldrich
Poly-L-proline, mol wt 1,000-10,000