Merck
Immunopontentiating and antitumor activities of a polysaccharide from Pulsatilla chinensis (Bunge) Regel.
International journal of biological macromolecules (2012-12-19)
Tong Liu, Leiguang Ye, Xueqing Guan, Xiaoshuan Liang, Cong Li, Qian Sun, Yang Liu, Shiru Chen, Fa Bang, Baogang Liu
ABSTRAKT
One water-soluble polysaccharide (PCPw) was isolated and purified from the roots of Pulsatilla chinensis by DEAE cellulose-52 and Sephadex G-100 column chromatography, and its antitumor activity was evaluated on 4T1 tumor-bearing mice through transplantable animal tumor. After 10 days of PCPw (50, 100 and 200 mg/kg) treatment once daily in tumor-bearing mice, PCPw oral administration could not only significantly inhibit the growth of transplantable 4T1 tumor in mice but also promote concanavalin A (Con A), lipopolysaccharide (LPS)-stimulated splenocytes proliferation, the serum lysozyme level and 2,4-dinitrofluorobenzene (DNFB)-induced delayed-type hypersensitivity (DTH) reactions, especially at the dose of 100 mg/kg. Meanwhile, significant improvements in peripheral blood abnormality and anemia were observed in PCPw-treated group. These results suggested that PCPw could improve both cellular and humoral immune response and might be explored as a potential natural antitumor drug.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Fluoro-2,4-dinitrobenzene, ≥99%
Supelco
1-Fluoro-2,4-dinitrobenzene, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
1-Fluoro-2,4-dinitrobenzene, purum p.a., ≥98.0% (GC)