Merck
Ascorbyl palmitate-loaded chitosan nanoparticles: characteristic and polyphenol oxidase inhibitory activity.
Colloids and surfaces. B, Biointerfaces (2012-12-19)
Mi Kyung Kim, Ji-Soo Lee, Kwang Yup Kim, Hyeon Gyu Lee
ABSTRAKT
The aim of this study was to produce ascorbyl palmitate (AP)-loaded nanoparticles in order to inhibit polyphenol oxidase (PPO) in bananas. AP-loaded chitosan nanoparticles were prepared using acetic acid and citric acid (denoted as CS/AA and CS/CA nanoparticles, respectively). As the initial AP concentration increases, the particle size significantly decreases, and the zeta potential, entrapment and loading efficiency significantly increases. The PPO inhibitory activity of AP was effectively improved when AP was nano-encapsulated by chitosan compared to no encapsulation. These results suggest that chitosan nano-encapsulation can be used to enhance the PPO inhibitory activity of AP.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ascorbic acid 6-palmitate, meets USP testing specifications
Sigma-Aldrich
6-O-Palmitoyl-L-ascorbic acid, BioXtra, ≥99.0% (RT)