Merck
[Topical treatment of hyperpigmentation disorders].
Annales de dermatologie et de venereologie (2012-12-25)
M Rendon, S Horwitz
ABSTRAKT
Hyperpigmentation has traditionally been a relatively difficult condition to treat, especially in darker racial ethnic groups. Multiple topical agents available act upon different steps of the pigmentation pathway. We review these topical agents, their mechanisms of action, and their effectiveness as monotherapy and in combination with other compounds. Ultimately, combination therapy is the most efficacious when considering overall depigmentation as well as treatment time required to achieve clinical improvement.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Supelco
Hydroquinone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland