Merck
[Effect of phenformin hydrochloride on pharmacokinetics of puerarin in rats].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-12-29)
Ying Deng, Ning Li, Mei Cui, Zhi-li Xiong, Fa-mei Li
ABSTRAKT
To study the effect of phenformin hydrochloride that may be illegally added in traditional Chinese medicine preparations on the pharmacokinetics of puerarin in rats. Rats were randomly divided into the single pueraria group and the phenformin hydrochloride combined with pueraria group. After oral administration in the two groups, their bloods were sampled at different time points to determine the drug concentration of puerarin in rat blood and calculate pharmacokinetic parameters. After oral administration with pueraria extracts and phenformin hydrochloride combined with pueraria extracts, the two groups showed main pharmacokinetic parameters as follows: Cmax were (2.39 +/- 1.01), (1.03 +/- 0.35) mg x L(-1), respectively; Tmax were (0.50 +/- 0.09), (1.5 +/- 0.5) h, respectively; Ke were (0.153 +/- 0.028), (0.172 +/- 0.042) h(-1), respectively; t(1/2) were (4.65 +/- 0.86), (4.20 +/- 0.81) h, respectively; AUC(0-t), were (5.73 +/- 2.60), (5.45 +/- 1.81) mg x h x L(-1), respectively; AUC(0-infinity) were (6.72 +/- 2.89), (6.26 +/- 1.88) mg x h x L(-1), respectively. Compared with the single puerarin group, the Cmax was significantly decreased (P < 0.05) and the Tmax was markedly longer (P < 0.01) than the hydrochloride combined with pueraria group. Phenformin hydrochloride can slow down the absorption process of puerarin and change the pharmacokinetic process of puerarin to some extent.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Puerarin, analytical standard
Sigma-Aldrich
Puerarin, ≥98.0% (HPLC)