Merck
Highly diastereoselective multicomponent cascade reactions: efficient synthesis of functionalized 1-indanols.
Angewandte Chemie (International ed. in English) (2013-01-03)
Jun Jiang, Xiaoyu Guan, Shunying Liu, Baiyan Ren, Xiaochu Ma, Xin Guo, Fengping Lv, Xiang Wu, Wenhao Hu
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,3-Benzofuran, 99%