Merck
Palladium-catalyzed oxidative cycloaddition through C-H/N-H activation: access to benzazepines.
Angewandte Chemie (International ed. in English) (2013-01-03)
Lei Wang, Jiayao Huang, Shiyong Peng, Hui Liu, Xuefeng Jiang, Jian Wang
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isatin, 97%