Merck
Comparison of superficial mycosis treatment using Butenafine and Bifonazole nitrate clinical efficacy.
Global journal of health science (2013-01-04)
Mohammed A Abdul Bari
ABSTRAKT
Superficial fungal infections are commonly encountered by the physician. And the continuously changing epidemiology of invasive fungal infections results in the need for an expanded armamentarium of antifungal therapies. This study was designed to evaluate the safety and efficacy of Butenafine (BTF) versus Bifonazole (BFZ) in the treatment of superficial mycosis in a randomized, double-blind, parallel-group trial. Of 96 patients, 48 applied (BTF) cream and 48 applied (BFZ) cream for 2 weeks to tinea versicolor, corporis and cruris treat, while tinea of feet & hands was treated for 4 weeks duration. Efficacy was assessed after the end of treatment and 2 weeks later. At the end of therapy, we find somewhat more patients using (BTF) than using (BFZ) had a mycologic cure ((BTF), 87.5%; (BFZ) 83.3%) and effective clinical response ((BTF), 91.7%; (BFZ), 83.3%). (BTF) provides rapid and persistent antifungal activity and symptom relief in patients with superficial mycosis during treatment. And patients continued to improve for at least 2 weeks after treatment. The Rates of mycologic cure and effective treatment with (BTF) were higher than with (BFZ) at cessation of treatment and 2 weeks later. However, no significant difference found between the two drugs (p> 0.05).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bifonazole, ≥98% (HPLC)
Bifonazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Bifonazole, Pharmaceutical Secondary Standard; Certified Reference Material