Merck
Revealing dynamic processes of materials in liquids using liquid cell transmission electron microscopy.
Journal of visualized experiments : JoVE (2013-01-05)
Kai-Yang Niu, Hong-Gang Liao, Haimei Zheng
ABSTRAKT
The recent development for in situ transmission electron microscopy, which allows imaging through liquids with high spatial resolution, has attracted significant interests across the research fields of materials science, physics, chemistry and biology. The key enabling technology is a liquid cell. We fabricate liquid cells with thin viewing windows through a sequential microfabrication process, including silicon nitride membrane deposition, photolithographic patterning, wafer etching, cell bonding, etc. A liquid cell with the dimensions of a regular TEM grid can fit in any standard TEM sample holder. About 100 nanoliters reaction solution is loaded into the reservoirs and about 30 picoliters liquid is drawn into the viewing windows by capillary force. Subsequently, the cell is sealed and loaded into a microscope for in situ imaging. Inside the TEM, the electron beam goes through the thin liquid layer sandwiched between two silicon nitride membranes. Dynamic processes of nanoparticles in liquids, such as nucleation and growth of nanocrystals, diffusion and assembly of nanoparticles, etc., have been imaged in real time with sub-nanometer resolution. We have also applied this method to other research areas, e.g., imaging proteins in water. Liquid cell TEM is poised to play a major role in revealing dynamic processes of materials in their working environments. It may also bring high impact in the study of biological processes in their native environment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pentadecane, ≥98.0% (GC)
Supelco
Pentadecane, analytical standard
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Pentadecane, ≥99%