- The use of brilliant green in ion-pair chromatography.
The use of brilliant green in ion-pair chromatography.
Talanta (1979-07-01)
J Dinunzio, H Freiser
PMID18962494
ABSTRAKT
Brilliant Green is used as the stationary phase in the ion-pair chromatography of butanoic, pentanoic, octanoic and lauric acids, with hexane-dichloromethane as eluent.