- Phasing out the use of stavudine in Singapore: how are we doing in compliance to World Health Organization (WHO) recommendations?
Phasing out the use of stavudine in Singapore: how are we doing in compliance to World Health Organization (WHO) recommendations?
Annals of the Academy of Medicine, Singapore (2013-01-11)
Mar Kyaw Win, Yee Sin Leo, Arlene Chua
PMID23303121
MATERIAŁY