Merck
Graphene oxide coated capillary for chiral separation by CE.
Electrophoresis (2013-01-12)
Nengsheng Ye, Jian Li, Yali Xie, Chang Liu
ABSTRAKT
This article describes a new application of graphene oxide (GO) in CE based on the coating of fused silica capillary for chiral separation. The coated capillary was characterized by SEM, energy dispersive X-ray spectroscopy, and Raman spectra. The results indicated that the capillary was coated with GO. Chiral separations were carried out in the GO-coated capillary for the ephedrine-pseudoephedrine (E-PE) isomers and β-methylphenethylamine (β-Me-PEA) isomers at the optimal buffer conditions without any chiral selector by CE. The precision and reproducibility of GO-coated capillary were investigated, and the RSDs of migration time (n = 6) for the E-PE isomers were 1.35-1.41%, and 0.97-3.50% for β-Me-PEA isomers, respectively. The LOD for E-PE isomers and β-Me-PEA isomers was 3 μM and 18 μM, respectively.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(1S,2S)-(+)-Pseudoephedrine, 98%
Sigma-Aldrich
(1R,2S)-(−)-Ephedrine, 98%